Order Entry
United States
ContactUsLinkComponent
1-(2-Deoxy-2-fluoroarabinofuranosyl)-uracil, [5-3H] ≥97% (by HPLC), MORpure™
Irritant
1-(2-Deoxy-2-fluoroarabinofuranosyl)-uracil, [5-3H] ≥97% (by HPLC), MORpure™
Catalog # 101972-602
CAS Number:  
undefined
Irritant
1-(2-Deoxy-2-fluoroarabinofuranosyl)-uracil, [5-3H] ≥97% (by HPLC), MORpure™
Catalog # 101972-602
Supplier Number:  MT1778-250UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    250 µCi
  • Cat. No.:
    101972-602
  • Supplier no.:
    MT1778-250UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:2087934-27-6