Order Entry
United States
ContactUsLinkComponent
p-Fluorophenol, [14C(U)] ≥97% (by HPLC)
Irritant
p-Fluorophenol, [14C(U)] ≥97% (by HPLC)
Catalog # 101971-946
CAS Number:  
undefined
Irritant
p-Fluorophenol, [14C(U)] ≥97% (by HPLC)
Catalog # 101971-946
Supplier Number:  MC460-100UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    100 µCi
  • Cat. No.:
    101971-946
  • Supplier no.:
    MC460-100UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:2088008-47-1