
Uridine-5'-triphosphate, tetraammonium salt, [2-14C] ≥95% (by HPLC)
Catalog # 101971-920
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:NextGen V-Vial
- Size:10 µCi
- Cat. No.:101971-920
- Supplier no.:MC450-10UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:289623-61-6