Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ioxynil octanoate, [ring-14C(U)]
Ioxynil octanoate, [ring-14C(U)] ≥98% (by HPLC), MORpure™
Ioxynil octanoate, [ring-14C(U)] ≥98% (by HPLC), MORpure™
Ioxynil octanoate, [ring-14C(U)] ≥98% (by HPLC), MORpure™
101971-464
:
MORAVEK BIOCHEMICALS MS
:
Ioxynil octanoate, [ring-14C(U)] ≥98% (by HPLC), MORpure™
101971-464
:
MORAVEK BIOCHEMICALS MS
:
MC2349-10UCI
:
Pack type:
NextGen V-Vial
Size:
10 µCi
Cat. No.:
101971-464
Supplier no.:
MC2349-10UCI
:
:
2910
:
,
:
GHS07
:
2088858-58-4