3,5-Diacetylaniline, [ring-14C(U)] ≥97% (by HPLC)
Catalog # 101971-064
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:Vial, amber
- Size:50 µCi
- Cat. No.:101971-064
- Supplier no.:MC2144-50UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:2088617-14-3