
P-Aminobenzoic acid [ring-14C] ≥98% (by HPLC), MORpure™
Catalog # 101970-446
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:NextGen V-Vial
- Size:50 µCi
- Cat. No.:101970-446
- Supplier no.:MC1391-50UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:912261-54-2