Order Entry
United States
ContactUsLinkComponent
3'-Azido-3'-deoxythymidine
3'-Azido-3'-deoxythymidine
Catalog # 101970-006
CAS Number:  
undefined
3'-Azido-3'-deoxythymidine
Catalog # 101970-006
Supplier Number:  M817-25MG
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial, with screwcap
  • Size:
    25 mg
  • Cat. No.:
    101970-006
  • Supplier no.:
    M817-25MG

Specifications

About this item

 :
 :267.25 g/mol
CAS Number:30516-87-1
Formula:C₁₀H₁₃N₅O₄
MDL Number:MFCD00006536