Order Entry
United States
ContactUsLinkComponent
2-Chloro-2'-deoxyadenosine-5'-monophosphate, diammonium salt, [8-14C] 100 µCi/ml in ethanol 50% , 98% (by HPLC), MORpure™, HPLC purified
2-Chloro-2'-deoxyadenosine-5'-monophosphate, diammonium salt, [8-14C] 100 µCi/ml in ethanol 50% , 98% (by HPLC), MORpure™, HPLC purified
Catalog # 101970-550
CAS Number:  
undefined
2-Chloro-2'-deoxyadenosine-5'-monophosphate, diammonium salt, [8-14C] 100 µCi/ml in ethanol 50% , 98% (by HPLC), MORpure™, HPLC purified
Catalog # 101970-550
Supplier Number:  MC1429-10UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    10 µCi
  • Cat. No.:
    101970-550
  • Supplier no.:
    MC1429-10UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:2089103-97-7