SP Bel-Art FLUO-KEM® Teflon® Resin Lab-Thread Tape, Bel-Art Products, a part of SP
Catalog # 490013-160
Supplier: Bel-Art Products, a Part of SP
Specifications
- Width:12.7 mm
- Color:White
- Description:Lab-thread tape
- Cat. No.:490013-160
- Supplier no.:F24020-0000
Specifications
About this item
FLUO-KEM resin lab-thread tape are pure, non-sintered contains no adhesives, eliminates the need for lubricants and vaccum greases.
- Provides a vacuum tight seal and eliminates freezing even after long periods
- Used on ground glass joints, desiccators, bell jars, freezedrying equipment, etc