Flash Point Reference Standards, Cannon Instrument Company
Supplier: Cannon
Certificates
About this item
Flash point reference materials are produced in accordance with ASTM D56, Tag closed cup flash point, ASTM D92, Cleveland open cup flash point and ASTM D93, Pensky Marten closed cup flash point
Specific data including average flash point and observed standard deviation (from a statistical analysis of 40 tests in 20 diff erent laboratories) is provided with each standard.
Frequently Bought Together