Perfringens Agar Base (O.P.S.P.) Dehydrated Culture Media, HiMedia Laboratories
Catalog # 95022-022
Supplier: HiMedia
CAS Number:
Specifications
- Description:Perfringens Agar Base (O.P.S.P.)
- Cat. No.:95022-022
- Supplier no.:M579-500G
Specifications
About this item
For the selective isolation and enumeration of Clostridium perfringens from foods.
- Microbiological culture media
- 500 g poly bottle