Sabourauds Chloramphenicol Agar, Modified, Dehydrated Culture Media, Himedia
Catalog # 95039-326
Supplier: HiMedia
CAS Number:
Specifications
- Description:Sabourauds Chloramphenicol Agar, Modified
- Cat. No.:95039-326
- Supplier no.:M1681-500G
Specifications
About this item
A selective medium recommended for the isolation of all species of yeast and dermatophytes.
- Microbiological culture media
- 500 g poly bottle