Salt Broth, Modified, Dehydrated Culture Media, Himedia
Catalog # 95020-412
Supplier: HiMedia
CAS Number:
Specifications
- Description:Salt Broth, Modified
- Cat. No.:95020-412
- Supplier no.:M1068-500G
Specifications
About this item
For the differentiation of enterococcal Group D streptococci from nonenterococcal Group D streptococci.
- Microbiological culture media
- 500 g poly bottle