Temp-Tec NL-517, Knitted Neoprene Gloves, Mapa Professionnel
: Avantor
Gloves are chemical-resistant, provide thermal insulation and extended length for extra arm protection.
- Insulated neoprene gloves
- Positive non-slip roughed grip
- Two-color construction for a visual indicator of glove wear-through
Gloves offer reliable hand protection for chemical processing, metal finishing, road works, polymer extrusion, Fish filleting and mould handling in ceramic industry.
: In compliance with 21 CFR 170-199.
Frequently Bought Together