D-(+)-Trehalose dihydrate NF, Ph. Eur. for bioprocessing, suitable for use as excipient endotoxin tested
Supplier: Avantor
Certificates
About this item
CAS Number:6138-23-4
:
Formula:C₁₂H₂₂O₁₁·2H₂O
MDL Number:MFCD00071594
:378.33 g/mol
Melting Pt:97…99 °C
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (NF- Anhydrous basis, EP- Anhydrous substance):97.0- 102.0%
- Appearance of solution (2.2.1 & 2.2.2):PASS
- Bacterial Endotoxins:≤ 2.5 EU/g
- Chloride<221> :≤ 0.0125%
- Chlorides (2.4.4):≤ 125 ppm
- Color and Clarity of Solution:PASS
- Escherichia coli <62>:ABSENT
- Heavy Metals <231>:≤ 5 ppm
- Identification A (USP, EP):PASS
- Identification B (USP, EP):PASS
- Identification C (USP, EP):PASS
- Microbial Contamination (2.6.12): TAMC:≤ 1000 CFU/g
- Microbial Contamination (2.6.12): TYMC:≤ 100 CFU/g
- Microbial Contamination (2.6.13): Escherichia coli:ABSENT
- Microbial Contamination (2.6.13): Salmonella:ABSENT
- Nitrogen Determination <461>:≤ 0.005%
- pH <791>, (2.2.3):4.5 - 6.5
- Related Substances (NF):≤ 0.5%
- Related Substances (NF- Disregard Limit):≤ 0.1%
- Related Substances (EP- Impurities A,B):≤ 0.5%
- Related Substances (EP- Total):≤ 1.0%
- Related Substances (EP- Unspecified Impurities):≤ 0.2%
- Residue after Ignition <281>:≤ 0.1%
- Salmonella <62>:ABSENT
- Soluble Starch Test (NF, EP):PASS
- Specific Rotation <781S>, (2.2.7):+197 - +201 Degrees
- Sulfated Ash (2.4.14):≤ 0.1%
- Sulfate <221>:≤ 0.0200%
- Sulfates (2.4.13):≤ 200 ppm
- Total Aerobic Microbial Count <61>:≤ 100 CFU/g
- Total Combined Molds and Yeasts Count <61>:≤ 100 CFU/g
- Water Determination <921>, (2.5.12):9.0 – 11.0%
Specification Test Results
Related Information
Certificates
Search by Lot Number25609588.pdf
SDS
spec_VWRB3599-12KG spec_VWRB3599-12KG spec_VWRB3599-12KG spec_VWRB3599-12KG
Product specification
VWR Bioprocessing Chemical and Excipients
Product Literature
Frequently Bought Together