Accessories for Chromatography Columns with Reservoir, Chemglass
Catalog # 80066-748
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Adapter Flow Control, [ST] Joints, 24/40
- Cat. No.:80066-748
- Supplier no.:CG-1191-01
Specifications
About this item
Flow control adapter for use with CG-1188 and CG-1189 chromatography columns along with CG-1190 reservoirs.
- 0 to 4 mm Chem-Cap® metering plug