ContactUsLinkComponent
BD Taxo™ Novobiocin Discs, BD Diagnostic Systems
BD Taxo™ Novobiocin Discs, BD Diagnostic Systems
Catalog # 90002-578
Supplier:  BD
CAS Number:  
BD Taxo™ Novobiocin Discs, BD Diagnostic Systems
Catalog # 90002-578
Supplier:  BD
Supplier Number:  231750
CAS Number:  
Specifications
  • Description:
    BD Taxo™ - Novobiocin Discs
  • Size:
    50 discs
  • Cat. No.:
    90002-578
  • Supplier no.:
    231750
Specifications
About this item
Taxo™ Differentiation Discs Novobiocin are recommended for the differentiation of coagulase negative staphylococci (e.g., Staphylococcus saprophyticus) based on novobiocin resistance.