ContactUsLinkComponent
Bacillus cereus Selective Supplement, EMD Millipore
Bacillus cereus Selective Supplement, EMD Millipore
Catalog # EM1.09875.0010
Supplier:  MilliporeSigma
CAS Number:  
Bacillus cereus Selective Supplement, EMD Millipore
Catalog # EM1.09875.0010
Supplier:  MilliporeSigma
Supplier Number:  1.09875.0010
CAS Number:  
Specifications
  • Pack type:
    Vial
  • Description:
    Bacillus cereus selective supplement
  • Size:
    10 x 1
  • Cat. No.:
    EM1.09875.0010
  • Supplier no.:
    1.09875.0010
Specifications
About this item
This Bacillus cereus Selective Supplement contains polymyxin B sulfate in lyophilized form.