Order Entry
United States
ContactUsLinkComponent
 
E.Z.N.A.® FFPE DNA Kit, Omega
 
E.Z.N.A.® FFPE DNA Kit, Omega

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    FFPE tissues
  • Target molecule:
    DNA
  • Description:
    E.Z.N.A.® FFPE DNA Kit

 

 

Frequently Bought Together