Order Entry
United States
ContactUsLinkComponent
Anion Calibration Standard (7 components) Inorganic Ventures
Anion Calibration Standard (7 components) Inorganic Ventures
Catalog # 103255-004
Supplier:  Inorganic Ventures
CAS Number:  
Anion Calibration Standard (7 components) Inorganic Ventures
Catalog # 103255-004
Supplier:  Inorganic Ventures
Supplier Number:  IV-STOCK-59-125
CAS Number:  

Specifications

  • Concentration:
    1 000 µg/ml in water
  • Description:
    Anion Calibration Standard, 7 components
  • Size:
    125 ml
  • Cat. No.:
    103255-004
  • Supplier no.:
    IVSTOCK-59-125

Specifications

About this item

This anion calibration standard is a certified reference material set in a water matrix for stability.

1 000 µg/mL ea: bromide, chloride, fluoride, nitrate, nitrite, o-phosphate, sulfate.

Certifications: ISO 17025 and ISO 17034 certified.