ContactUsLinkComponent
Storage Canes for Cryovial® Tubes, Simport
Storage Canes for Cryovial® Tubes, Simport
Catalog # 89511-292
Supplier:  Simport Scientific
undefined
Storage Canes for Cryovial® Tubes, Simport
Catalog # 89511-292
Supplier:  Simport Scientific
Supplier Number:  T313
Specifications
  • Description:
    Cane for Cryovial® tubes, aluminium
  • Cat. No.:
    89511-292
  • Supplier no.:
    T313
Specifications
About this item
These 290 mm long storage canes for Cryovial® tubes are made of aluminium and can be used for storage of up to five 1.2 or 2 mL tubes in liquid nitrogen containers.