iScript™ Reverse Transcription Supermix for RT-qPCR for fast synthesis of first-strand cDNA for direct use in real-time qPCR
- Combined oligo(dT) and random primers provide unbiased representation of 5' and 3' regions
- Contains RNase H+ MMLV reverse transcriptase and has a linear range of 1 µg – 1 pg total input RNA
- Amplify cDNA for genes with low expression levels
- Fast cDNA synthesis protocol
- 50 reactions of no-RT controls for optional detection of gDNA amplicon contamination
Storage temperature: Store at –20°C.
This single-tube supermix contains all the components for reverse transcription except RNA template. It is a sensitive, fast and convenient reagent for gene expression analysis.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.