M-Tergitol 7 Agar Base Dehydrated Culture Media, HiMedia
Catalog # 95020-406
Supplier: HiMedia
CAS Number:
Specifications
- Description:M-Tergitol 7 Agar Base
- Cat. No.:95020-406
- Supplier no.:M1066-500G
Specifications
About this item
For the selective isolation and identification of injured coliforms from chlorinated water using membrane filter technique.
- Microbiological culture media
- 500 g poly bottles