10X TBS Wash Buffer, with Tween 20, Sera Care
Supplier: KPL
Certificates
About this item
10X Tris-buffered Saline (TBS) with Tween 20 is used as a common western blot wash solution and general wash buffer in immunoassay applications
Formulation of TBST with higher concentration of Tween 20 provides a more stringent wash than standard TBST formulations.
Tween 20, a non-ionic detergent, reduces non-specific binding and protein-protein interaction during the wash step in immunoassay procedures.