Order Entry
United States
Orders LinkContactUsLinkComponent
 

Sequencng Binding Buffer, Zymo Research

Supplier:   Zymo Research
Sequencng Binding Buffer, Zymo Research
Supplier:  Zymo Research
Certificates
Sequencng Binding Buffer, Zymo Research
Sequencng Binding Buffer, Zymo Research

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 500 mL
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $456.39
    $456.39
    Each (500ml)
       $456.39
     

    Supplier Number: D4050-1-500
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 55 mL
    Log in for availability
    Log in for availability
    Each (55ml)
    Each (55ml) - $116.23
    $116.23
    Each (55ml)
       $116.23
     

    Supplier Number: D4050-1-55
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.