Anti-PD-L1 (B7-H1) mouse Monoclonal Antibody [clone: 29E.2A3]
Supplier: Bio X Cell
Certificates
About this item
InVivoMab anti-human PD-L1 (B7-H1), Clone: 29E.2A3, Isotype: mouse IgG2b, κ, Applications: in vitro PD-L1 blockade, Functional assays, Immunohistochemistry (frozen), Flow cytometry
Type: Primary
Antigen: PD-L1 (B7-H1)
Clonality: Monoclonal
Clone: 29E.2A3
Conjugation:
Epitope:
Host: mouse
Isotype: IgG2b
Reactivity: Human
Specifications
- Catalog No:
- 76556-990
- 76555-874
- 76554-604
- 76555-988
- 76555-382
- Antigen Symbol:
- PD-L1
- PD-L1
- PD-L1
- PD-L1
- PD-L1
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 29E.2A3
- 29E.2A3
- 29E.2A3
- 29E.2A3
- 29E.2A3
- Reactivity:
- Human
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- IgG2b
- IgG2b
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
Specifications
Frequently Bought Together