About this item
InVivoPlus anti-mouse Ly6G, Clone: 1A8, Isotype: rat IgG2a, κ, Applications: in vivo neutrophil depletion, in vivo MDSC depletion, Immunofluorescence, Immunohistochemistry (paraffin), Immunohistochemistry (frozen), Flow cytometry
Type: Primary
Antigen: Ly6G
Clonality: Monoclonal
Clone: 1A8
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76554-880
- 76554-994
- 76555-450
- 76554-934
- Antigen Symbol:
- Ly6G
- Ly6G
- Ly6G
- Ly6G
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 1A8
- 1A8
- 1A8
- 1A8
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Rat
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes