Order Entry
United States
ContactUsLinkComponent
 
Anti-MHC Class I (H-2Kb) bound to SIINFEKL peptide (OVA residues 257-264) Mouse Monoclonal Antibody [clone: 25-D1.16]
 
Anti-MHC Class I (H-2Kb) bound to SIINFEKL peptide (OVA residues 257-264) Mouse Monoclonal Antibody [clone: 25-D1.16]

 

  • Catalog No:
  • 76555-254
  • 76556-204
  • 76556-028
  • 76554-494
  • 76554-626
  • Antigen Symbol:
  • MHC Class I
  • MHC Class I
  • MHC Class I
  • MHC
  • MHC Class I
  • Antigen Name:
  • MHC Class I (H-2Kb) bound to SIINFEKL peptide (OVA residues 257-264)
  • MHC Class I (H-2Kb) bound to SIINFEKL peptide (OVA residues 257-264)
  • MHC Class I (H-2Kb) bound to SIINFEKL peptide (OVA residues 257-264)
  • MHC Class I (H-2Kb) bound to SIINFEKL peptide (OVA residues 257-264)
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • 25-D1.16
  • 25-D1.16
  • 25-D1.16
  • 25-D1.16
  • 25-D1.16
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Yes

 

Frequently Bought Together