Anti-Vbeta4 TCR Rat Monoclonal Antibody [clone: KT4]
InVivoMab anti-mouse Vβ4 TCR, Clone: KT4, Isotype: rat IgG2b, Applications: in vivo administration, Flow cytometry
Type: Primary
Antigen: Vbeta4 TCR
Clonality: Monoclonal
Clone: KT4
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
- Catalog No:
- 76555-058
- 76555-036
- 76557-148
- 76554-942
- 76555-046
- Antigen Symbol:
- Vbeta4 TCR
- Vbeta4 TCR
- Vbeta4 TCR
- Vbeta4 TCR
- Vbeta4 TCR
- Antigen Name:
- Vbeta4 TCR
- Vbeta4 TCR
- Vbeta4 TCR
- Vbeta4 TCR
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- KT4
- KT4
- KT4
- KT4
- KT4
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Rat
- Rat
- Isotype:
- IgG2b
- IgG2b
- IgG2b
- IgG2b
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
Frequently Bought Together