Order Entry
United States
ContactUsLinkComponent
Anti-MHC Class I (H-2Kb) Mouse Monoclonal Antibody [clone: AF6-88.5.5.3]
Anti-MHC Class I (H-2Kb) Mouse Monoclonal Antibody [clone: AF6-88.5.5.3]
Supplier:  Bio X Cell
Certificates
Anti-MHC Class I (H-2Kb) Mouse Monoclonal Antibody [clone: AF6-88.5.5.3]
Supplier:  Bio X Cell

Specifications

  • Catalog No:
  • 76554-396
  • 76555-564
  • 76556-298
  • 76555-206
  • 76555-778
  • Antigen Symbol:
  • MHC Class I (H-2Kb)
  • MHC Class I (H-2Kb)
  • MHC
  • MHC Class I (H-2Kb)
  • MHC Class I (H-2Kb)
  • Antigen Name:
  • MHC Class I (H-2Kb)
  • MHC Class I (H-2Kb)
  • MHC Class I (H-2Kb)
  • MHC Class I (H-2Kb)
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • AF6-88.5.5.3
  • AF6-88.5.5.3
  • AF6-88.5.5.3
  • AF6-88.5.5.3
  • AF6-88.5.5.3
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a
  • IgG2a
  • IgG2a
  • IgG2a
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Yes

Specifications

Frequently Bought Together