InVivoMab anti-mouse TIM-3 (CD366), Clone: RMT3-23, Isotype: rat IgG2a, κ, Applications: in vivo TIM-3 neutralization, in vitro TIM-3 blocking, Flow cytometry
Type: Primary
Antigen: TIM-3 (CD366)
Clonality: Monoclonal
Clone: RMT3-23
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76556-376
- 76554-420
- 76554-816
- 76555-520
- 76555-768
- Antigen Symbol:
- TIM-3
- TIM-3
- TIM-3
- TIM-3
- TIM-3
- Antigen Name:
- TIM-3 (CD366)
- TIM-3 (CD366)
- TIM-3 (CD366)
- TIM-3 (CD366)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- RMT3-23
- RMT3-23
- RMT3-23
- RMT3-23
- RMT3-23
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Rat
- Rat
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Yes
- Yes