Anti-MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7) Mouse Monoclonal Antibody [clone: 10-3.6.2]
Supplier: Bio X Cell
Certificates
About this item
InVivoMab anti-mouse MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7), Clone: 10-3.6.2, Isotype: mouse IgG2a, Applications: in vitro MHC class II I-A blocking, in vitro MHC class II I-A expressing cell negative selection
Type: Primary
Antigen: MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7)
Clonality: Monoclonal
Clone: 10-3.6.2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Specifications
- Catalog No:
- 76554-812
- 76555-216
- 76556-320
- 76556-520
- 76556-394
- Antigen Symbol:
- MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7)
- MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7)
- MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7)
- MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7)
- MHC
- Antigen Name:
- MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7)
- MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7)
- MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7)
- MHC Class II (I-Ak, I-Ar, I-Af, I-As,I-Ag7)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 10-3.6.2
- 10-3.6.2
- 10-3.6.2
- 10-3.6.2
- 10-3.6.2
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
Specifications
Frequently Bought Together