Order Entry
United States
ContactUsLinkComponent
Anti-Thy1.2 (CD90.2) Rat Monoclonal Antibody [clone: 30H12]
Anti-Thy1.2 (CD90.2) Rat Monoclonal Antibody [clone: 30H12]
Supplier:  Bio X Cell
Certificates
Anti-Thy1.2 (CD90.2) Rat Monoclonal Antibody [clone: 30H12]
Supplier:  Bio X Cell

Specifications

  • Catalog No:
  • 76556-792
  • 76554-400
  • 76556-972
  • 76555-576
  • 76554-678
  • Antigen Symbol:
  • Thy1.2
  • Thy1.2 (CD90.2)
  • Thy1.2 (CD90.2)
  • Thy1.2 (CD90.2)
  • Thy1.2 (CD90.2)
  • Antigen Name:
  • Thy1.2 (CD90.2)
  • Thy1.2 (CD90.2)
  • Thy1.2 (CD90.2)
  • Thy1.2 (CD90.2)
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • 30H12
  • 30H12
  • 30H12
  • 30H12
  • 30H12
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Rat
  • Rat
  • Isotype:
  • IgG2b
  • IgG2b
  • IgG2b
  • IgG2b

Specifications

Frequently Bought Together