Anti-MHC Class I (H-2Kk, H-2Dk) Mouse Monoclonal Antibody [clone: 16-1-2N (HB14)]
Supplier: Bio X Cell
Certificates
About this item
InVivoMab anti-mouse MHC Class I (H-2Kk, H-2Dk), Clone: 16-1-2N (HB14), Isotype: mouse IgG2a, Applications: Functional assays, Flow cytometry
Type: Primary
Antigen: MHC Class I (H-2Kk, H-2Dk)
Clonality: Monoclonal
Clone: 16-1-2N (HB14)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Specifications
- Catalog No:
- 76556-110
- 76555-942
- 76554-776
- 76554-362
- 76555-588
- Antigen Symbol:
- MHC Class I (H-2Kk, H-2Dk)
- MHC
- MHC Class I (H-2Kk, H-2Dk)
- MHC Class I (H-2Kk, H-2Dk)
- MHC Class I (H-2Kk, H-2Dk)
- Antigen Name:
- MHC Class I (H-2Kk, H-2Dk)
- MHC Class I (H-2Kk, H-2Dk)
- MHC Class I (H-2Kk, H-2Dk)
- MHC Class I (H-2Kk, H-2Dk)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 16-1-2N (HB14)
- 16-1-2N (HB14)
- 16-1-2N (HB14)
- 16-1-2N (HB14)
- 16-1-2N (HB14)
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
Specifications
Frequently Bought Together