Anti-CD8beta (Lyt 3.2) Rat Monoclonal Antibody [clone: 53-5.8]
Supplier: Bio X Cell
Certificates
About this item
InVivoMab anti-mouse CD8β (Lyt 3.2), Clone: 53-5.8, Isotype: rat IgG1, κ, Applications: in vivo CD8+ T cell depletion, in vitro CD8 blockade, Immunofluorescence
Type: Primary
Antigen: CD8beta (Lyt 3.2)
Clonality: Monoclonal
Clone: 53-5.8
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Specifications
- Catalog No:
- 76556-400
- 76554-710
- 76556-214
- 76555-420
- 76556-476
- Antigen Symbol:
- CD8beta (Lyt 3.2)
- CD8beta
- CD8beta (Lyt 3.2)
- CD8beta (Lyt 3.2)
- CD8beta (Lyt 3.2)
- Antigen Name:
- CD8beta (Lyt 3.2)
- CD8beta (Lyt 3.2)
- CD8beta (Lyt 3.2)
- CD8beta (Lyt 3.2)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 53-5.8
- 53-5.8
- 53-5.8
- 53-5.8
- 53-5.8
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Rat
- Rat
- Isotype:
- IgG1
- IgG1
- IgG1
- IgG1
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes
Specifications
Frequently Bought Together