Anti-TYRP1/TRP1 (gp75) Mouse Monoclonal Antibody [clone: TA99]
Supplier: Bio X Cell
Certificates
About this item
InVivoMab anti-mouse/human TYRP1/TRP1 (gp75), Clone: TA99, Isotype: mouse IgG2a, κ, Applications: in vivo triggering of FcγRs, Immunofluorescence, Flow cytometry
Type: Primary
Antigen: TYRP1/TRP1 (gp75)
Clonality: Monoclonal
Clone: TA99
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Human
Specifications
- Catalog No:
- 76555-758
- 76556-534
- 76555-598
- 76555-030
- 76555-952
- Antigen Symbol:
- TYRP1/TRP1 (gp75)
- TYRP1/TRP1
- TYRP1/TRP1 (gp75)
- TYRP1/TRP1 (gp75)
- TYRP1/TRP1 (gp75)
- Antigen Name:
- mouse/TYRP1/TRP1 (gp75)
- mouse/TYRP1/TRP1 (gp75)
- mouse/TYRP1/TRP1 (gp75)
- mouse/TYRP1/TRP1 (gp75)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- TA99
- TA99
- TA99
- TA99
- TA99
- Reactivity:
- Human, Mouse
- Human, Mouse
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
Specifications
Frequently Bought Together