Order Entry
United States
Orders LinkContactUsLinkComponent
Benzonase®

Benzonase®

Benzonase®
Supplier:  MilliporeSigma
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Suitable for biopharmaceutical production EMPROVE® bio.

  • Activity (DNA; pH 8.0; 30 min; 37 °C) : ≥250
  • U/µL
  • Proteases: not detectable
  • Endotoxines (LAL-test): <0.25 EU in 1000 U
  • Colony count (aerobic bacteria), (Yeasts and moulds): <10 CFU in 100 000 U, <10 CFU in 100 000 U
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • EM1.01654.0001
  • EM1.01695.0001
  • EM1.01656.0001
  • EM1.01697.0001
  • EM1.01697.0010
  • Enzyme Name:
  • Benzonase®
  • Benzonase®
  • Benzonase®
  • Benzonase®
  • Benzonase®
  • Form:
  • Colorless
  • Colorless
  • Colorless
  • Colorless
  • Colorless
  • Purity:
  • ≥99.0%(calc. based on protein), Grade II
  • ≥99.0%(calc. based on protein), Grade I
  • ≥99.0%(calc. based on protein), Grade II
  • ≥99.0%(calc. based on protein), Grade I
  • ≥99.0%(calc. based on protein), Grade I
  • Storage Conditions:
  • Store at −20 °C. Transport with dry ice
  • Store at −20 °C. Transport with dry ice
  • Store at −20 °C. Transport with dry ice
  • Store at −20 °C. Transport with dry ice
  • Store at −20 °C. Transport with dry ice
  • Enzyme Activity:
  • ≥1.1E+06 U/mg
  • ≥1.1E+06 U/mg
  • ≥1.0E+06 U/mg
  • ≥1.1E+06 U/mg
  • ≥1.1E+06 U/mg

Specifications

Product Family Options

Product Information

  • Size
    Pack type
    Availability
    Price
  • 100000 U
    Plastic bottle
    Discontinued
    Supplier Number: 1.01654.0001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    EM1.01654.0001
    Enzyme Name
    Benzonase®
    Form
    Colorless
    Purity
    ≥99.0%(calc. based on protein), Grade II
    Storage Conditions
    Store at −20 °C. Transport with dry ice
    Enzyme Activity
    ≥1.1E+06 U/mg
  • 100000 U
    Plastic bottle
    Discontinued
    Supplier Number: 1.01695.0001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    EM1.01695.0001
    Enzyme Name
    Benzonase®
    Form
    Colorless
    Purity
    ≥99.0%(calc. based on protein), Grade I
    Storage Conditions
    Store at −20 °C. Transport with dry ice
    Enzyme Activity
    ≥1.1E+06 U/mg
  • 500000 U
    Aluminium tube
    Discontinued
    Supplier Number: 1.01656.0001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    EM1.01656.0001
    Enzyme Name
    Benzonase®
    Form
    Colorless
    Purity
    ≥99.0%(calc. based on protein), Grade II
    Storage Conditions
    Store at −20 °C. Transport with dry ice
    Enzyme Activity
    ≥1.0E+06 U/mg
  • 500000 U
    Aluminium tube
    Log in for availability
    Log in for availability
    Each
    Each - $11,198.60
    $11,198.60
    Each
       $11,198.60

    Supplier Number: 1.01697.0001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    EM1.01697.0001
    Enzyme Name
    Benzonase®
    Form
    Colorless
    Purity
    ≥99.0%(calc. based on protein), Grade I
    Storage Conditions
    Store at −20 °C. Transport with dry ice
    Enzyme Activity
    ≥1.1E+06 U/mg
  • 5000000 U
    Plastic bottle
    Log in for availability
    Log in for availability
    Each
    Each

    Supplier Number: 1.01697.0010
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    EM1.01697.0010
    Enzyme Name
    Benzonase®
    Form
    Colorless
    Purity
    ≥99.0%(calc. based on protein), Grade I
    Storage Conditions
    Store at −20 °C. Transport with dry ice
    Enzyme Activity
    ≥1.1E+06 U/mg

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.