Rat IgG1 Isotype Control [clone:TNP6A7]
Supplier: Bio X Cell
Certificates
About this item
InVivoPlus rat IgG1 isotype control, Clone: TNP6A7, Isotype: rat IgG1, λ, Applications: in vivo blocking of PD-1/PD-L signaling, in vitro PD-1 neutralization, Immunohistochemistry (frozen), Flow cytometry, Western blot
- Recommended Dilution Buffer: IP0070
Type: Control
Antigen:
Clonality: Monoclonal
Clone: TNP6A7
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- 76555-232
- 76556-330
- 76555-824
- 76556-980
- Antibody Type:
- Control
- Control
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- TNP6A7
- TNP6A7
- TNP6A7
- TNP6A7
- Host:
- Rat
- Rat
- Rat
- Rat
- Isotype:
- IgG1
- IgG1
- IgG1
- IgG1
Specifications
Frequently Bought Together