Mouse IgG2b Isotype Control [clone:MPC-11]
InVivoPlus mouse IgG2b isotype control, Clone: MPC-11, Isotype: mouse IgG2b, κ, Applications: in vivo administration, Functional assays, Immunohistochemistry (frozen), Immunohistochemistry (paraffin), Western blot, Immunofluorescence, Flow Cytometry
- Recommended Dilution Buffer: IP0070
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MPC-11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
- Catalog No:
- 76556-556
- 76556-486
- 76554-850
- 76555-068
- Antibody Type:
- Control
- Control
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- MPC-11
- MPC-11
- MPC-11
- MPC-11
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- IgG2b
- IgG2b
- Western Blot:
- Yes
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
Frequently Bought Together