Order Entry
United States
ContactUsLinkComponent
Rat IgG1 Anti-HRP Isotype Control [clone: HRPN]
Rat IgG1 Anti-HRP Isotype Control [clone: HRPN]
Supplier:  Bio X Cell
Certificates
Rat IgG1 Anti-HRP Isotype Control [clone: HRPN]
Supplier:  Bio X Cell

Specifications

  • Catalog No:
  • 76557-276
  • 76555-368
  • 76555-794
  • 76557-074
  • 76556-590
  • Antigen Symbol:
  • HRP
  • Rat
  • HRP
  • HRP
  • HRP
  • Antigen Name:
  • Horseradish Peroxidase
  • Horseradish Peroxidase
  • Horseradish Peroxidase
  • Horseradish Peroxidase
  • Antibody Type:
  • Control
  • Control
  • Control
  • Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • HRPN
  • HRPN
  • HRPN
  • HRPN
  • HRPN
  • Host:
  • Rat
  • Rat
  • Rat
  • Rat
  • Rat
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • IgG1

Specifications

Frequently Bought Together