Anti-PRELP Rabbit Polyclonal Antibody
Catalog # 76072-426
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Prolargin
- Antigen Symbol:PRELP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:119-148 aa
- Form:Liquid
- Gene ID:5549
- Modification:None
- Molecular Weight:44 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This PRELP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 119-148 amino acids from the Central region of human PRELP.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76072-426
Specifications
About this item
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100
Type: Primary
Antigen: PRELP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 119-148
Host: Rabbit
Isotype: Ig
Reactivity: Human