Anti-IL24 Rabbit Polyclonal Antibody
400 μL
Catalog # 76072-136
Supplier: Prosci
Supplier #: 62-096
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
For WB starting dilution is: 1:1000
Type: Primary
Antigen: IL24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 139-168
Host: Rabbit
Isotype: Ig
Reactivity: Human
Specifications
- Size:400 μL
- Clonality:Polyclonal
- Modification:None
- Molecular Weight:24 kDa
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Epitope:139-168 aa
- Form:Liquid
- Host:Rabbit
- Concentration:0.5 mg/ml
- Cat. No.:76072-136
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4°C
- Antibody Type:Primary
- Antigen Symbol:IL24
- Gene ID:11009
- Immunogen:This IL24 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 139-168 amino acids from the C-terminal region of human IL24.
- Conjugation:Unconjugated
- Antigen Name:Interleukin-24
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



