Anti-SLC13A5 Rabbit Polyclonal Antibody
Catalog # 76071-260
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Solute carrier family 13 member 5
- Antigen Symbol:SLC13A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 μL
- Epitope:151-180 aa
- Form:Liquid
- Gene ID:284111
- Modification:None
- Molecular Weight:63 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This S13A5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 151-180 amino acids from the Central region of human S13A5.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76071-260
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: SLC13A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 151-180
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse