Anti-LRRC74B Rabbit Polyclonal Antibody
76064-772
: Prosci
- Antibody Type:Primary
- Antigen Name:Leucine-rich repeat-containing protein 74B {ECO:0000312|HGNC:HGNC:34301}
- Antigen Symbol:LRRC74B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:355-383 aa
- Form:Liquid
- Gene ID:400891
- Modification:None
- Molecular Weight:42 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This YV012 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 355-383 amino acids from the C-terminal region of human YV012.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76064-772
For WB starting dilution is: 1:1000
Type: Primary
Antigen: LRRC74B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 355-383
Host: Rabbit
Isotype: Ig
Reactivity: Human