Anti-MED19 Rabbit Polyclonal Antibody
Catalog # 76062-744
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Mediator of RNA polymerase II transcription subunit 19
- Antigen Symbol:MED19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:161-188 aa
- Form:Liquid
- Gene ID:219541
- Modification:None
- Molecular Weight:26 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.45 mg/ml
- Shipping Temperature:4°C
- Immunogen:This MED19 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 161-188 amino acids from the C-terminal region of human MED19.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76062-744
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: MED19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 161-188
Host: Rabbit
Isotype: Ig
Reactivity: Human