Anti-ARSD Rabbit Polyclonal Antibody
400 μL
Catalog # 76066-676
Supplier: Prosci
Supplier #: 58-635
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: ARSD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 502-532
Host: Rabbit
Isotype: Ig
Reactivity: Human
Specifications
- Size:400 μL
- Clonality:Polyclonal
- Modification:None
- Molecular Weight:65 kDa
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Epitope:502-532 aa
- Form:Liquid
- Host:Rabbit
- Concentration:0.5 mg/ml
- Cat. No.:76066-676
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4°C
- Antibody Type:Primary
- Antigen Symbol:ARSD
- Gene ID:414
- Immunogen:This ARSD antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 502-532 amino acids from the C-terminal region of human ARSD.
- Conjugation:Unconjugated
- Antigen Name:Arylsulfatase D
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



