Anti-Stk25 Rabbit Polyclonal Antibody
76067-988
: Prosci
- Antibody Type:Primary
- Antigen Name:Serine/threonine-protein kinase 25
- Antigen Symbol:Stk25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:327-355 aa
- Form:Liquid
- Gene ID:59041
- Modification:None
- Molecular Weight:48 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This Mouse Stk25 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 327-355 amino acids from the C-terminal region of mouse Stk25.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76067-988
For WB starting dilution is: 1:1000
Type: Primary
Antigen: Stk25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 327-355
Host: Rabbit
Isotype: Ig
Reactivity: Human