Anti-HDAC4 Rabbit Polyclonal Antibody
Catalog # 76067-134
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Histone deacetylase 4
- Antigen Symbol:HDAC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:1049-1077 aa
- Form:Liquid
- Gene ID:9759
- Modification:None
- Molecular Weight:119 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.35 mg/ml
- Shipping Temperature:4°C
- Immunogen:This HDAC4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1049-1077 amino acids from the C-terminal region of human HDAC4.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76067-134
Specifications
About this item
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50 For IF starting dilution is: 1:10~50
Type: Primary
Antigen: HDAC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 1049-1077
Host: Rabbit
Isotype: Ig
Reactivity: Human