OIML Class F2 Precision Weight Sets, Mettler Toledo
Basic Line weights are protected in robust and easy to clean plastic boxes
- Made of stainless steel
- Adjustable cavity (AC) design
- Economical
FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
: All weight sets delivered with tweezers.
Frequently Bought Together