Glucose Yeast Extract Agar Dehydrated Culture Media, HiMedia
Catalog # 95022-542
Supplier: HiMedia
CAS Number:
Specifications
- Description:Glucose Yeast Extract Agar
- Cat. No.:95022-542
- Supplier no.:M963-500G
Specifications
About this item
For the enumeration and cultivation of lactobacilli in pharmaceutical preparations.
- Microbiological culture media
- 500 g poly bottles